Learn More
Selleck Chemical LLC PRI-724 5mg
SDP
Supplier:  Selleck Chemical LLC S89685MG
Encompass_Preferred
PRI-724 (C-82 prodrug, ICG-001 analog) is a potent and specific inhibitor that disrupts the interaction of ?-catenin and CBP. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3460