Learn More
Aviva Systems Biology Corp PRICKLE3 Antibody - C-terminal region (ARP34413_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34413_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRICKLE3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PRICKLE3. Alternate names: LMO6ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-52954