Learn More
Medchemexpress LLC Pridinol mesylate | 6856-31-1 | 100.0% | 100 MG
SDP
Catalog No. 5000412069
Encompass_Preferred
Pridinol mesylate is an orally active, blood-brain permeable, muscarinic acetylcholine receptor (mAChR)-directed muscle relaxant. It reduces the conduction of impulses to spinal motor neurons and exerts muscle relaxant activity.
  • Inhibits skeletal muscle contractures in diseases of both central and peripheral origin.
  • Can be used in research in the field of musculoskeletal diseases.
Catalog No. 50-004-12069 Supplier Medchemexpress LLC Supplier No. HYB1343100MG
Pridinol mesylate is an orally active, blood-brain permeable, muscarinic acetylcholine receptor (mAChR)-directed muscle relaxant. It reduces the conduction of impulses to spinal motor neurons and exerts muscle relaxant activity.
  • Inhibits skeletal muscle contractures in diseases of both central and peripheral origin.
  • Can be used in research in the field of musculoskeletal diseases.