Learn More
Medchemexpress LLC Pridinol mesylate | 6856-31-1 | MFCD00079241 | 99.97% | 50 MG
SDP
Catalog No. 5000411867
Encompass_Preferred
Pridinol mesylate is an orally active, blood-brain permeable muscle relaxant, targeting muscarinic acetylcholine receptors. It reduces impulses to spinal motor neurons, inhibiting skeletal muscle contractures of central and peripheral origin.
  • Muscarinic acetylcholine receptor-directed muscle relaxant
  • Reduces conduction of impulses to spinal motor neurons
  • Inhibits skeletal muscle contractures
  • Used in research for musculoskeletal diseases
Catalog No. 50-004-11867 Supplier Medchemexpress LLC Supplier No. HYB134350MG
Pridinol mesylate is an orally active, blood-brain permeable muscle relaxant, targeting muscarinic acetylcholine receptors. It reduces impulses to spinal motor neurons, inhibiting skeletal muscle contractures of central and peripheral origin.
  • Muscarinic acetylcholine receptor-directed muscle relaxant
  • Reduces conduction of impulses to spinal motor neurons
  • Inhibits skeletal muscle contractures
  • Used in research for musculoskeletal diseases