Learn More
Apexbio Technology LLC Pridinol Methanesulfonate 6856-31-1 5g
SDP
Supplier:  Apexbio Technology LLC A85075000
Encompass_Preferred
Pridinol Methanesulfonate (CAS 6856-31-1) is a synthetic small molecule classified as an anticholinergic compound It exerts its pharmacological effects primarily by inhibiting muscarinic acetylcholine receptors thereby reducing cholinergic neurotransmission within the central nervous system This mechanism underlies its application as an antiparkinsonian agent in biomedical research where it is utilized to study neuromuscular transmission and motor control disorders associated with altered cholinergic signaling Pridinol Methanesulfonate serves as a valuable tool for investigating the pathophysiology of movement disorders and evaluating novel therapeutics that target cholinergic pathways
Catalog No. 50-000-95175