Learn More
Medchemexpress LLC HY-10684 5mg Medchemexpress, Pridopidine CAS:346688-38-8 Purity:>98%
SDP
Catalog No. 501968356
Encompass_Preferred
Medchemexpress, HY-10684 5mg Pridopidine CAS:346688-38-8 Pridopidine, a dopamine (DA) stabilizer, acts as a low affinity dopamine D2 receptor (D2R) antagonist. Pridopidine exerts high affinity towards sigma 1 receptor (S1R) with Ki between 70 and 80 nM, which is ~100× higher than its affinity toward D2R. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8356 Supplier Medchemexpress LLC Supplier No. HY106845MG
Medchemexpress, HY-10684 5mg Pridopidine CAS:346688-38-8 Pridopidine, a dopamine (DA) stabilizer, acts as a low affinity dopamine D2 receptor (D2R) antagonist. Pridopidine exerts high affinity towards sigma 1 receptor (S1R) with Ki between 70 and 80 nM, which is ~100× higher than its affinity toward D2R. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.