Learn More
ATCC Priestia megaterium
Supplier:  ATCC 14581
Encompass
Priestia megaterium is a whole genome-sequenced bacterial type strain. This bacterium is listed as a quality control strain for the VITEK 2 BCL Comprehensive QC Set.
Catalog No. 50-238-4586