Learn More
Apexbio Technology LLC Prilocaine 721-50-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062041
Encompass_Preferred
Prilocaine (CAS 721-50-6) is a local anesthetic molecule categorized within the amino amide class Its primary pharmacological mechanism involves reversible blockade of voltage-gated sodium channels in neuronal membranes thereby inhibiting action potential propagation and sensory nerve impulse conduction Due to its sodium channel-blocking profile prilocaine is commonly investigated in biomedical research examining reversible neural inhibition pain modulation pathways and sensory signal transmission mechanisms
Catalog No. 50-000-62041 Supplier Apexbio Technology LLC Supplier No. B21195.1
Prilocaine (CAS 721-50-6) is a local anesthetic molecule categorized within the amino amide class Its primary pharmacological mechanism involves reversible blockade of voltage-gated sodium channels in neuronal membranes thereby inhibiting action potential propagation and sensory nerve impulse conduction Due to its sodium channel-blocking profile prilocaine is commonly investigated in biomedical research examining reversible neural inhibition pain modulation pathways and sensory signal transmission mechanisms