Learn More
Medchemexpress LLC HY-B0137 50mg Medchemexpress, Prilocaine CAS:721-50-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB013750MG
Encompass_Preferred
Medchemexpress, HY-B0137 50mg Prilocaine CAS:721-50-6 Prilocaine, an amino amide, can anaesthetize animals in local. Prilocaine is a Na, K-ATPase inhibitor. Prilocaine has neurotoxic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2378