Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prilocaine, 98%

Catalog No. AC456220010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456220010 1 g
AC456220050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456220010 Supplier Thermo Scientific Chemicals Supplier No. 456220010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 721-50-6 | C13H20N2O | 220.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 721-50-6
Molecular Formula C13H20N2O
Molecular Weight (g/mol) 220.32
MDL Number MFCD00048681
InChI Key MVFGUOIZUNYYSO-UHFFFAOYNA-N
Synonym prilocaine, propitocaine, citanest, prilocainum, prilocaina, astra 1515, n-2-methylphenyl-2-propylamino propanamide, o-methyl-2-propylaminopropionanilide, propanamide, n-2-methylphenyl-2-propylamino, astra 1512
PubChem CID 4906
ChEBI CHEBI:8404
IUPAC Name N-(2-methylphenyl)-2-(propylamino)propanamide
SMILES CCCNC(C)C(=O)NC1=CC=CC=C1C

Specifications

Melting Point 36.0°C to 38.7°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 220.31
Percent Purity 98%
Chemical Name or Material Prilocaine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
EINECSNumber 211-957-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.