Learn More
Medchemexpress LLC Prilocaine-d7 hydrochloride | 1786-81-8 | 98.2% | 100 UG
SDP
Catalog No. 5000450694
Encompass_Preferred
Prilocaine-d7 (hydrochloride) is a deuterium-labeled form of Prilocaine (hydrochloride). Prilocaine hydrochloride is an amino amide and a Na, K-ATPase inhibitor known for its neurotoxic effects. Stable heavy isotopes like deuterium are incorporated into drug molecules primarily as tracers for quantitation during drug development, with potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-50694 Supplier Medchemexpress LLC Supplier No. HYB0137AS100UG
Prilocaine-d7 (hydrochloride) is a deuterium-labeled form of Prilocaine (hydrochloride). Prilocaine hydrochloride is an amino amide and a Na, K-ATPase inhibitor known for its neurotoxic effects. Stable heavy isotopes like deuterium are incorporated into drug molecules primarily as tracers for quantitation during drug development, with potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.