Learn More
Medchemexpress LLC Prilocaine-d7 hydrochloride | 98.2% | 500 UG
SDP
Catalog No. 5000450915
Encompass_Preferred
Prilocaine-d7 hydrochloride is a deuterium-labeled form of Prilocaine hydrochloride, an amino amide and a Na, K-ATPase inhibitor. Known to have neurotoxic effects, this compound is for research use only and not sold to patients. Its deuterated nature can impact pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
Catalog No. 50-004-50915 Supplier Medchemexpress LLC Supplier No. HYB0137AS500UG
Prilocaine-d7 hydrochloride is a deuterium-labeled form of Prilocaine hydrochloride, an amino amide and a Na, K-ATPase inhibitor. Known to have neurotoxic effects, this compound is for research use only and not sold to patients. Its deuterated nature can impact pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS