Learn More
Selleck Chemical LLC Prilocaine 10mg 721-50-6 NSC 40027
SDP
Supplier:  Selleck Chemical LLC S161910MG
Encompass_Preferred
Prilocaine (NSC 40027) is a local anesthetic of the amino amide type. It acts on sodium channels on the neuronal cell membrane, limiting the spread of seizure activity and reducing seizure propagation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-771-5