Learn More
Medchemexpress LLC Prilocaine hydrochloride | 1786-81-8 | 100 MG
SDP
Catalog No. 5000450770
Encompass_Preferred
Prilocaine hydrochloride is an amino amide that functions as a Na/K-ATPase inhibitor, known to exhibit neurotoxic effects. It is intended for research and analytical applications.
  • Amino amide and Na/K-ATPase inhibitor
  • Exhibits neurotoxic effects
  • Intended for research and analytical applications
  • Inhibits Na,K-ATPase of plasma membranes of LM cells with IC50 values of 43.8 mM at 37 °C and 28.2 mM at 25 °C
Catalog No. 50-004-50770 Supplier Medchemexpress LLC Supplier No. HYB0137A100MG
Prilocaine hydrochloride is an amino amide that functions as a Na/K-ATPase inhibitor, known to exhibit neurotoxic effects. It is intended for research and analytical applications.
  • Amino amide and Na/K-ATPase inhibitor
  • Exhibits neurotoxic effects
  • Intended for research and analytical applications
  • Inhibits Na,K-ATPase of plasma membranes of LM cells with IC50 values of 43.8 mM at 37 °C and 28.2 mM at 25 °C