Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prilocaine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18605809 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-809 25 g Poly Bottle
18-605-810 100 g Poly Bottle
18-605-811 500 g Poly Pail
18-605-812 1 kg Poly Pail
18-605-813 5 kg Poly Pail
18-605-808 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-605-809 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PR10225GM
Add to Cart
Add to Cart

PR102, 1786-81-8, C13H20N2O·HCl

Spectrum™ Chemical Prilocaine Hydrochloride, USP is a local anesthetic used most often for infiltration anesthesia in dentistry. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1786-81-8
Molecular Formula C13H21ClN2O
Molecular Weight (g/mol) 256.77
InChI Key BJPJNTKRKALCPP-UHFFFAOYNA-N
IUPAC Name hydrogen N-(2-methylphenyl)-2-(propylamino)propanamide chloride
SMILES [H+].[Cl-].CCCNC(C)C(=O)NC1=CC=CC=C1C

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.003
Packaging Poly Bottle
Quantity 25 g
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.