Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prilocaine, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18605819 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-819 5 kg Amber Glass Bottle
18-605-814 5 g Amber Glass Bottle
18-605-815 25 g Amber Glass Bottle
18-605-816 100 g Amber Glass Bottle
18-605-817 500 g Amber Glass Bottle
18-605-818 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-605-819 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P19545KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

P1954, 721-50-6, C16H22N2O3

Spectrum™ Chemical Prilocaine, USP is a local anesthetic used most often for infiltration anesthesia in dentistry. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 721-50-6
Molecular Formula C13H20N2O
Molecular Weight (g/mol) 220.32
MDL Number MFCD00048681
InChI Key MVFGUOIZUNYYSO-UHFFFAOYNA-N
IUPAC Name N-(2-methylphenyl)-2-(propylamino)propanamide
SMILES CCCNC(C)C(=O)NC1=CC=CC=C1C

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 5 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Prilocaine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.