Learn More
Apexbio Technology LLC Primary Antibody Dilution Buffer 100mL
SDP
Catalog No. 503107205
Encompass_Preferred
This buffer mainly consists of BSA Triton-100 etc which helps antibody penetration into tissues and blocks non-specific signals It is primarily used for diluting and preparing primary antibodies
Catalog No. 50-310-7205 Supplier Apexbio Technology LLC Supplier No. K1200100
This buffer mainly consists of BSA Triton-100 etc which helps antibody penetration into tissues and blocks non-specific signals It is primarily used for diluting and preparing primary antibodies