Learn More
Apexbio Technology LLC Primary Antibody Dilution Buffer 500mL
SDP
Catalog No. 503107139
Encompass_Preferred
This buffer mainly consists of BSA Triton-100 etc which helps antibody penetration into tissues and blocks non-specific signals It is primarily used for diluting and preparing primary antibodies
Catalog No. 50-310-7139 Supplier Apexbio Technology LLC Supplier No. K1200500
This buffer mainly consists of BSA Triton-100 etc which helps antibody penetration into tissues and blocks non-specific signals It is primarily used for diluting and preparing primary antibodies