Learn More
Lonza Walkersville P4 Primary Cell 4D-Nucleofector X Kit S
Catalog No. NC1022582
Encompass
The P4 Primary Cell 4D-Nucleofector X Kit S is one of our five 4D-Nucleofector Kits suited for transfecting primary cells in the 20 µL 16-well Nucleocuvette Strip format when working with the 4D-Nucleofector X Unit.Benefits:High transfection efficiencies for primary cellsCells maintain functionality post transfectionConditions are transferable between 4D-Nucleofector System, 96-well Shuttle System and 384-well Nucleofector SystemApplications:Transfection of low cell numbers in the range of 5 x 10⁴ to 1 x 10⁶ cellsMedium throughput applicationsOptimization of transfection conditions e.g. substrate titrationsContent:1 x 0.675 mL P4 Primary Cell Nucleofector Solution1 x 0.15 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)2 x 16-well Nucleocuvette Strips (20 μl)
Catalog No. NC1022582 Supplier Lonza Walkersville Supplier No. V4XP4032
The P4 Primary Cell 4D-Nucleofector X Kit S is one of our five 4D-Nucleofector Kits suited for transfecting primary cells in the 20 µL 16-well Nucleocuvette Strip format when working with the 4D-Nucleofector X Unit.Benefits:High transfection efficiencies for primary cellsCells maintain functionality post transfectionConditions are transferable between 4D-Nucleofector System, 96-well Shuttle System and 384-well Nucleofector SystemApplications:Transfection of low cell numbers in the range of 5 x 10⁴ to 1 x 10⁶ cellsMedium throughput applicationsOptimization of transfection conditions e.g. substrate titrationsContent:1 x 0.675 mL P4 Primary Cell Nucleofector Solution1 x 0.15 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)2 x 16-well Nucleocuvette Strips (20 μl)