Learn More
Cayman Chemical Prim-O-Glucosylcimifugin, Purity: ≥98%, CAS: 80681-45-4, 30 mg
SDP
Catalog No. 502910730
Encompass_Preferred
A chromone with diverse biological activities; inhibits proliferation of EGF-stimulated MCF-7 cells at 10 µM; reduces LPS-induced increases in the production of NO, TNF-α, IL-6, and IL-1β in RAW 264.7 macrophages at 15, 50, and 100 µg/ml; decreases pulmonary edema in a mouse model of LPS-induced acute lung injury at 2.5-10 mg/kg; inhibits formalin-induced tonic pain in rats (ED50 = 1.6 mg/animal)
Catalog No. 50-291-0730 Supplier Cayman Chemical Supplier No. 3492130 MG
A chromone with diverse biological activities; inhibits proliferation of EGF-stimulated MCF-7 cells at 10 µM; reduces LPS-induced increases in the production of NO, TNF-α, IL-6, and IL-1β in RAW 264.7 macrophages at 15, 50, and 100 µg/ml; decreases pulmonary edema in a mouse model of LPS-induced acute lung injury at 2.5-10 mg/kg; inhibits formalin-induced tonic pain in rats (ED50 = 1.6 mg/animal)