Learn More
Medchemexpress LLC HY-N0635 5mg Medchemexpress, Prim-O-glucosylcimifugin CAS:80681-45-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06355MG
Encompass_Preferred
Medchemexpress, HY-N0635 5mg Prim-O-glucosylcimifugin CAS:80681-45-4 Prim-O-glucosylcimifugin exerts anti-inflammatory effects through the inhibition of iNOS and COX-2 expression by through regulating JAK2/STAT3 signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3650