Learn More
Aviva Systems Biology Corp Prim1 Antibody - C-terminal region (ARP46014_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46014_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Prim1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI324982, MGC107288ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-236