Learn More
Selleck Chemical LLC PRIMA-1-10mg
SDP
Catalog No. 502389942
Encompass_Preferred
PRIMA-1 (2,2-Bis(hydroxymethyl)-3-quinuclidinone) is a mutant p53 reactivator. It induces apoptosis and inhibits growth of human tumors with mutant p53.
Catalog No. 50-238-9942 Supplier Selleck Chemical LLC Supplier No. S772310MG
PRIMA-1 (2,2-Bis(hydroxymethyl)-3-quinuclidinone) is a mutant p53 reactivator. It induces apoptosis and inhibits growth of human tumors with mutant p53.