Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prima-1 | 5608-24-2 | 98.0% | 185.22 | 200 MG
SDP

Catalog No. 5000405076
Encompass_Preferred

PRIMA-1 (NSC-281668) is a mutant p53 reactivator that restores the sensitivity of TP53 mutant-type thyroid cancer cells to the histone methylation inhibitor 3-Deazaneplanocin A.

  • Purity of 98.0%
  • Solid appearance
  • White to off-white color
  • Soluble in H2O and DMSO
  • Stable for up to 3 years as powder and 2 years in solvent when stored correctly

Catalog No. 50-004-05076 Supplier Medchemexpress LLC Supplier No. HY19980A200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PRIMA-1 (NSC-281668) is a mutant p53 reactivator that restores the sensitivity of TP53 mutant-type thyroid cancer cells to the histone methylation inhibitor 3-Deazaneplanocin A.

  • Purity of 98.0%
  • Solid appearance
  • White to off-white color
  • Soluble in H2O and DMSO
  • Stable for up to 3 years as powder and 2 years in solvent when stored correctly

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.