Learn More
Enzo Life Sciences PRIMA-1 (5mg). CAS: 5608-24-2
SDP
Supplier:  Enzo Life Sciences BMLGR3330005
Encompass_Preferred
Novel cell-permeable p53 reactivating agent. Restores sequence-specific DNA binding and the active conformation to mutant p53 (in 13 of 14 p53 mutants tested) in vitro and in living cells. Induces apoptosis in human tumor cell lines and displays antitumor activity in mice. Alternative name: 2,2-bis(Hydroxymethyl)-1-azabicyclo[2.2.2]octan-3-one, 2,2-bis(Hydroxymethyl)-3-quinuclidinone. Purity: ≥98% (TLC). Solubility: Soluble at 25 mg/ml in ethanol, 100% DMSO, and DMF. Soluble at 10 mg/ml in water. Long Term Storage: -20°C.
Catalog No. 50-201-0988