Learn More
Tocris Bioscience™ PRIMA-1MET

Description
Biological Activity: Methylated derivative of PRIMA-1. Restores mutant activity p53 activity. Acts synergistically with chemotherapeutic agents to inhibit tumor cell proliferation.
Storage: Store at 4°C
Specifications
Specifications
| Description | CAS: 5291-32-7 |
| Formulation | C10H17NO3 |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 10 mg |
| Product Type | Inhibitors |
| Molecular Weight (g/mol) | 199.25g/mol |
| Solubility | Soluble to 100mM in DMSO, 100mM in ethanol |
| Content And Storage | 4°C |
| Target | PRIMA-1MET |
| Color | Dark Purple |
For Laboratory Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.