Learn More
Medchemexpress LLC HY-19980 10mg Medchemexpress, PRIMA-1Met CAS:5291-32-7 Purity:>98%
SDP
Catalog No. 502029303
Encompass_Preferred
Medchemexpress, HY-19980 10mg PRIMA-1Met CAS:5291-32-7 PRIMA-1Met restores wild-type conformation and function to mutant p53, and triggers apoptosis in tumor cells. PRIMA-1Met also targets the selenoprotein thioredoxin reductase 1 (TrxR1), a key regulator of cellular redox balance. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9303 Supplier Medchemexpress LLC Supplier No. HY1998010MG
Add to Cart
Add to Cart
Medchemexpress, HY-19980 10mg PRIMA-1Met CAS:5291-32-7 PRIMA-1Met restores wild-type conformation and function to mutant p53, and triggers apoptosis in tumor cells. PRIMA-1Met also targets the selenoprotein thioredoxin reductase 1 (TrxR1), a key regulator of cellular redox balance. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.