Learn More
Pion Inc PRIMA HT 50ML
Catalog No. NC0914983
Encompass
Proprietary buffer for performing PAMPA or µSOL assays. Quantity: 1 bottle; volume: 50 mL per bottle.
Catalog No. NC0914983 Supplier Pion Inc Supplier No. 110151
Proprietary buffer for performing PAMPA or µSOL assays. Quantity: 1 bottle; volume: 50 mL per bottle.