Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amsbio LLC (formerly Genlantis) PrimaPure™ Human Endothelial Cell Growth Media

Catalog No. PM112500
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Human Microvascular Endothelial Cells (HMVEC)
Without Additives:
None
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Without Additives
PM112500 Human Microvascular Endothelial Cells (HMVEC) None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PM112500 Supplier Amsbio LLC (formerly Genlantis) Supplier No. AMS.PM112500
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimized for endothelial cells

Specifications

Product Type Endothelial Cell Growth Media
Without Additives None
For Use With (Application) Human Microvascular Endothelial Cells (HMVEC)
Quantity 500 mL
Content And Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.