Learn More
Apexbio Technology LLC Primaquine Diphosphate 63-45-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062489
Encompass_Preferred
Primaquine Diphosphate is an 8-aminoquinoline derivative utilized primarily as an antimalarial agent targeting the extraerythrocytic liver stage of malaria parasites While its precise IC50 values remain unspecified primaquine s biological activity involves interference with nucleic acid function subsequently disrupting protein synthesis pathways and lipid biosynthesis and inducing membrane disturbances In vitro primaquine has been applied in cell culture models to investigate antiviral effects demonstrating dose-dependent inhibition of Newcastle disease virus replication and modulation of viral RNA synthesis In vivo experiments using radiolabeled primaquine-loaded liposomes in murine models illustrate pronounced hepatic accumulation Clinically this compound is employed as prophylaxis against malaria infections (e g Plasmodium falciparum and P vivax)
Catalog No. 50-000-62489 Supplier Apexbio Technology LLC Supplier No. B18205.1
Primaquine Diphosphate is an 8-aminoquinoline derivative utilized primarily as an antimalarial agent targeting the extraerythrocytic liver stage of malaria parasites While its precise IC50 values remain unspecified primaquine s biological activity involves interference with nucleic acid function subsequently disrupting protein synthesis pathways and lipid biosynthesis and inducing membrane disturbances In vitro primaquine has been applied in cell culture models to investigate antiviral effects demonstrating dose-dependent inhibition of Newcastle disease virus replication and modulation of viral RNA synthesis In vivo experiments using radiolabeled primaquine-loaded liposomes in murine models illustrate pronounced hepatic accumulation Clinically this compound is employed as prophylaxis against malaria infections (e g Plasmodium falciparum and P vivax)