Learn More
Selleck Chemical LLC Primaquine Diphosphate S4237-1g
SDP
Catalog No. 503108638
Encompass_Preferred
Primaquine Diphosphate is a transmission-blocking anti-malarial clinically available displaying a marked activity against gametocytes of all species of human malaria
Catalog No. 50-310-8638 Supplier Selleck Chemical LLC Supplier No. S42371G
Primaquine Diphosphate is a transmission-blocking anti-malarial clinically available displaying a marked activity against gametocytes of all species of human malaria