Learn More
Apexbio Technology LLC Primaquine Diphosphate,5g CAS# 63-45-6
SDP
Supplier:  Apexbio Technology LLC B18205000
Encompass_Preferred
Primaquine, an 8-aminoquinoline, is introduced as a curative antimalarial agent in 1950. Since then, the drug has been applied extensively to against the exoerythrocytic stage of malaria. It is demonstrated tthat primaquine, by binding to nucleic acids, could therefore block protein synthesis, alter lipid synthesis and interact with biological membranes. [1] Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0310