Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Primaquine diphosphate, 98%

Catalog No. ac170870050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC170870050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC170870050 Supplier Thermo Scientific Chemicals Supplier No. 170870050
Only null left
Add to Cart

CAS: 63-45-6 | C15H21N32H3PO4 | 455.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-45-6
Molecular Formula C15H21N32H3PO4
Molecular Weight (g/mol) 455.34
InChI Key GJOHLWZHWQUKAU-UHFFFAOYSA-N
Synonym primaquine phosphate, n4-6-methoxyquinolin-8-yl pentane-1,4-diamine phosphate, n-5-aminopentan-2-yl-6-methoxyquinolin-8-amine; phosphoric acid, 4-n-6-methoxyquinolin-8-yl pentane-1,4-diamine; phosphoric acid, primaquine diphosphate, chembl994, phosphoric acid; primaquine, 1, n4-6-methoxy-8-quinolinyl-, phosphate 1:2, n-5-aminopentan-2-yl-6-methoxyquinolin-8-amine phosphoric acid, n4-6-methoxyquinolin-8-yl pentane-1,4-diamine; phosphoric acid
PubChem CID 6135
IUPAC Name 4-N-(6-methoxyquinolin-8-yl)pentane-1,4-diamine;phosphoric acid
SMILES CC(CCCN)NC1=C2C(=CC(=C1)OC)C=CC=N2.OP(=O)(O)O.OP(=O)(O)O

Specifications

Melting Point 205.0°C to 206.0°C
Infrared Spectrum Authentic
Quantity 5 g
Merck Index 15,7863
Solubility Information Solubility in water: moderately soluble.
Formula Weight 455.34
Percent Purity 98%
Chemical Name or Material Primaquine diphosphate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 200-560-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.