Learn More
Biotium Primary Antibodies for Organelles Fluorescence Microscopy Super Resolution Imaging Cell Surface and Membrane Dyes 1/EA
Catalog No. 5000499391
Encompass_Preferred
CellBrite Steady Membrane dyes label surface membranes of live cells for 24 hours or longer The kits include CellBrite Steady Enhancer which allows selective imaging of plasma membrane fluorescence Red fluorescent CellBrite Steady 568 has Ex/Em at 569/586 nm 500 labelings
Catalog No. 50-004-99391 Supplier Biotium Supplier No. 30150
CellBrite Steady Membrane dyes label surface membranes of live cells for 24 hours or longer The kits include CellBrite Steady Enhancer which allows selective imaging of plasma membrane fluorescence Red fluorescent CellBrite Steady 568 has Ex/Em at 569/586 nm 500 labelings