Learn More
Biotium Primary Antibodies for Organelles Fluorescence Microscopy Super Resolution Imaging Cell Surface and Membrane Dyes 1/EA
Supplier:  Biotium 30150T
Encompass_Preferred
CellBrite Steady Membrane dyes label surface membranes of live cells for 24 hours or longer The kits include CellBrite Steady Enhancer which allows selective imaging of plasma membrane fluorescence Red fluorescent CellBrite Steady 568 has Ex/Em at 569/586 nm 100 labelings
Catalog No. 50-004-99693