Learn More
Biotium Primary Antibodies for Organelles Fluorescence Microscopy Super Resolution Imaging Cell Surface and Membrane Dyes 1/EA
Catalog No. 5000500835
Encompass_Preferred
CellBrite Steady Membrane dyes label surface membranes of live cells for 24 hours or longer The kits include CellBrite Steady Enhancer which allows selective imaging of plasma membrane fluorescence Near-IR fluorescent CellBrite Steady 750 has Ex/Em at 755/789 nm 100 labelings
Catalog No. 50-005-00835 Supplier Biotium Supplier No. 30141T
CellBrite Steady Membrane dyes label surface membranes of live cells for 24 hours or longer The kits include CellBrite Steady Enhancer which allows selective imaging of plasma membrane fluorescence Near-IR fluorescent CellBrite Steady 750 has Ex/Em at 755/789 nm 100 labelings