Learn More
Biotium Primary Antibodies for Organelles Fluorescence Microscopy Super Resolution Imaging Cell Surface and Membrane Dyes 1/EA
Catalog No. 5000500836
Encompass_Preferred
CellBrite Steady Membrane dyes label surface membranes of live cells for 24 hours or longer The kits include CellBrite Steady Enhancer which allows selective imaging of plasma membrane fluorescence Red fluorescent CellBrite Steady 594 has Ex/Em at 593/614 nm 500 labelings
Catalog No. 50-005-00836 Supplier Biotium Supplier No. 30142
CellBrite Steady Membrane dyes label surface membranes of live cells for 24 hours or longer The kits include CellBrite Steady Enhancer which allows selective imaging of plasma membrane fluorescence Red fluorescent CellBrite Steady 594 has Ex/Em at 593/614 nm 500 labelings