Learn More
Biotium Primary Antibody Beta 3 Tubulin r2G10 CF594 1/EA
Catalog No. 5000500714
Encompass_Preferred
Beta-3-Tubulin (r2G10) is a recombinant mouse monoclonal antibody that recognizes Beta-3-Tubulin This CF594 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00714 Supplier Biotium Supplier No. P047594500UL
Beta-3-Tubulin (r2G10) is a recombinant mouse monoclonal antibody that recognizes Beta-3-Tubulin This CF594 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability