Learn More
Biotium Primary Antibody Beta 3 Tubulin r2G10 CF640R 1/EA
Catalog No. 5000500723
Encompass_Preferred
Beta-3-Tubulin (r2G10) is a recombinant mouse monoclonal antibody that recognizes Beta-3-Tubulin This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00723 Supplier Biotium Supplier No. P047640R500UL
Add to Cart
Add to Cart
Beta-3-Tubulin (r2G10) is a recombinant mouse monoclonal antibody that recognizes Beta-3-Tubulin This CF640R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability