Learn More
Biotium Primary Antibody Biotin rBN-34 CF568 1/EA
Catalog No. 5000499972
Encompass_Preferred
Biotin (rBN-34) is a recombinant mouse monoclonal antibody that recognizes free biotin and biocytin This CF568 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99972 Supplier Biotium Supplier No. P0335681ML
Add to Cart
Add to Cart
Biotin (rBN-34) is a recombinant mouse monoclonal antibody that recognizes free biotin and biocytin This CF568 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability