Learn More
Biotium Primary Antibody Biotin rBN-34 CF680R 1/EA
Catalog No. 5000499463
Encompass_Preferred
Biotin (rBN-34) is a recombinant mouse monoclonal antibody that recognizes free biotin and biocytin This CF680R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99463 Supplier Biotium Supplier No. P033680R1ML
Biotin (rBN-34) is a recombinant mouse monoclonal antibody that recognizes free biotin and biocytin This CF680R antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability