Learn More
Biotium Primary Antibody c Fos r2H2 CF647 1/EA
Catalog No. 5000499997
Encompass_Preferred
c-Fos (r2H2) is a recombinant mouse monoclonal antibody that recognizes c-Fos This CF647 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99997 Supplier Biotium Supplier No. P043647500UL
c-Fos (r2H2) is a recombinant mouse monoclonal antibody that recognizes c-Fos This CF647 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability