Learn More
Biotium Primary Antibody c Fos r2H2 CF740 1/EA
Catalog No. 5000500605
Encompass_Preferred
c-Fos (r2H2) is a recombinant mouse monoclonal antibody that recognizes c-Fos This CF740 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00605 Supplier Biotium Supplier No. P043740500UL
c-Fos (r2H2) is a recombinant mouse monoclonal antibody that recognizes c-Fos This CF740 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability