Learn More
Biotium Primary Antibody CD19 SJ25C1 CF575 1/EA
Catalog No. 5000499027
Encompass_Preferred
CD19 (clone SJ25C1) is a mouse monoclonal antibody that recognizes the human protein CD19 CD19 is a common marker of B cells and is a B cell coreceptor This CF575 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-99027 Supplier Biotium Supplier No. P010575125
CD19 (clone SJ25C1) is a mouse monoclonal antibody that recognizes the human protein CD19 CD19 is a common marker of B cells and is a B cell coreceptor This CF575 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests