Learn More
Biotium Primary Antibody CD29 NA ExoBrite 410 450 1/EA
Supplier:  Biotium P040410500
Encompass_Preferred
ExoBrite 410/450 CD29 Flow Antibody is optimized for detection of CD29/Integrin beta-1 on isolated exosomes by flow cytometry The conjugate has Ex/Em at 408/452 nm for detection in the Pacific Blue channel Recommended for instruments optimized for small particle flow 100 tests
Catalog No. 50-005-00604