Learn More
Biotium Primary Antibody CD29 NA ExoBrite 410 450 1/EA
Catalog No. 5000500460
Encompass_Preferred
ExoBrite 410/450 CD29 Flow Antibody is optimized for detection of CD29/Integrin beta-1 on isolated exosomes by flow cytometry The conjugate has Ex/Em at 408/452 nm for detection in the Pacific Blue channel Recommended for instruments optimized for small particle flow 25 tests
Catalog No. 50-005-00460 Supplier Biotium Supplier No. P040410125
ExoBrite 410/450 CD29 Flow Antibody is optimized for detection of CD29/Integrin beta-1 on isolated exosomes by flow cytometry The conjugate has Ex/Em at 408/452 nm for detection in the Pacific Blue channel Recommended for instruments optimized for small particle flow 25 tests