Learn More
Biotium Primary Antibody CD29 NA ExoBrite 560 585 1/EA
Supplier:  Biotium P040560500
Encompass_Preferred
ExoBrite 560/585 CD29 Flow Antibody is optimized for detection of CD29/Integrin beta-1 on isolated exosomes by flow cytometry The conjugate has Ex/Em at 562/583 nm for detection in the PE channel off the yellow laser line Recommended for instruments optimized for small particle flow 100 tests
Catalog No. 50-004-99741