Learn More
Biotium Primary Antibody CD29 NA ExoBrite 640 660 1/EA
Supplier:  Biotium P040640500
Encompass_Preferred
ExoBrite 640/660 CD29 Flow Antibody is optimized for detection of CD29/Integrin beta-1 on isolated exosomes by flow cytometry The conjugate has Ex/Em at 640/660 nm Recommended for instruments optimized for small particle flow 100 tests
Catalog No. 50-005-00711