Learn More
Biotium Primary Antibody CD3e SK7 CF514 1/EA
Catalog No. 5000498510
Encompass_Preferred
CD3 (clone SK7) is a mouse monoclonal antibody that recognizes the human protein CD3 CD3 is a common marker of T cells and is a T cell coreceptor This CF514 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-98510 Supplier Biotium Supplier No. P002514125
CD3 (clone SK7) is a mouse monoclonal antibody that recognizes the human protein CD3 CD3 is a common marker of T cells and is a T cell coreceptor This CF514 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests