Learn More
Biotium Primary Antibody CD3e SK7 CF820 1/EA
Catalog No. 5000498151
Encompass_Preferred
CD3 (clone SK7) is a mouse monoclonal antibody that recognizes the human protein CD3 CD3 is a common marker of T cells and is a T cell coreceptor This CF820 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-98151 Supplier Biotium Supplier No. P002820125
CD3 (clone SK7) is a mouse monoclonal antibody that recognizes the human protein CD3 CD3 is a common marker of T cells and is a T cell coreceptor This CF820 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests